Gene Transcripts
- Gene Symbol
- TTC28-AS1
- Entrez Gene ID
- 284900
- Gene Name
- TTC28 antisense RNA 1
- Gene Aliases
- TTC28-AS, TTC28AS, dJ353E16.2
- Chromosome Location
- Chr. 22: 27919376 - 28002679 on Build GRCh38
- UniGene ID
- Hs.602319
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB051435.1 |
- |
1 |
726 |
|
CT841515.1 |
- |
1 |
267 |
|
DA934139.1 |
- |
4 |
466 |
|
View Full Product Details