Gene Transcripts

Gene Symbol
C3orf70
Entrez Gene ID
285382
Gene Name
chromosome 3 open reading frame 70
Gene Aliases
Chromosome Location
Chr. 3: 185078050 - 185153014 on Build GRCh38
UniGene ID
Hs.380048
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025266.1 NP_001020437.1 2 751
GenBank mRNA BC137178.1 - 2 618
BC137179.1 - 2 618 9021176
BC150565.1 - 2 629 40127634

View Full Product Details