Gene Transcripts

Gene Symbol
XKR6
Entrez Gene ID
286046
Gene Name
XK related 6
Gene Aliases
C8orf21, C8orf5, C8orf7, XRG6
Chromosome Location
Chr. 8: 10896144 - 11205789 on Build GRCh38
UniGene ID
Hs.657083
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173683.3 NP_775954.2 3 1027
GenBank mRNA AY534244.1 - 3 1027
BC024146.2 - 3 330 3678468

View Full Product Details