Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014553.2 | NP_055368.1 | 14, 15 | 1483 | |
XM_017003902.1 | XP_016859391.1 | 13, 14 | 1348 | ||
XM_017003903.1 | XP_016859392.1 | 9, 10 | 790 | ||
GenBank mRNA | AF198488.1 | - | 14, 15 | 1480 | |
AK311692.1 | - | 2, 3 | 113 | ||
BC064698.1 | - | 14, 15 | 1483 | 30347863 |