Gene Transcripts
- Gene Symbol
- HOXA4
- Entrez Gene ID
- 3201
- Gene Name
- homeobox A4
- Gene Aliases
- HOX1, HOX1D
- Chromosome Location
- Chr. 7: 27128507 - 27130780 on Build GRCh38
- UniGene ID
- Hs.533357
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002141.4 |
NP_002132.3
|
1 |
617 |
|
GenBank mRNA |
M74297.1 |
- |
1 |
570 |
|
View Full Product Details