Gene Transcripts

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
Chromosome Location
Chr. 11: 417930 - 442011 on Build GRCh38
UniGene ID
Hs.501622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012302.2 NP_001012302.2 7, 8 618
XM_011520048.1 XP_011518350.1 7, 8 618
XM_011520051.2 XP_011518353.1 7, 8 618
XM_011520052.2 XP_011518354.1 7, 8 618
XM_011520053.2 XP_011518355.1 7, 8 618
XM_017017644.1 XP_016873133.1 7, 8 618
XM_017017645.1 XP_016873134.1 7, 8 618
XM_017017646.1 XP_016873135.1 7, 8 618
XM_017017647.1 XP_016873136.1 7, 8 618
GenBank mRNA AK026923.1 - 7, 8 565
AK074088.1 - 4, 5 2031
AK303642.1 - 6, 7 660
AK316479.1 - 6, 7 686
BC128557.1 - 7, 8 586 40122929
BC144415.1 - 6, 7 644

View Full Product Details