Gene Transcripts

Gene Symbol
ATP13A5
Entrez Gene ID
344905
Gene Name
ATPase 13A5
Gene Aliases
Chromosome Location
Chr. 3: 193274789 - 193378824 on Build GRCh38
UniGene ID
Hs.658032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198505.2 NP_940907.2 6 588
XM_011512770.1 XP_011511072.1 6 687
XM_017006304.1 XP_016861793.1 4 230
GenBank mRNA AK122613.1 - 708
AY823163.1 - 6 588

View Full Product Details