Gene Transcripts

Gene Symbol
ITPKA
Entrez Gene ID
3706
Gene Name
inositol-trisphosphate 3-kinase A
Gene Aliases
IP3-3KA, IP3KA
Chromosome Location
Chr. 15: 41493858 - 41503559 on Build GRCh38
UniGene ID
Hs.2722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002220.2 NP_002211.1 2 592
XM_011521522.2 XP_011519824.1 3 273
GenBank mRNA BC026331.1 - 2 576
X54938.1 - 2 533

View Full Product Details