Gene Transcripts

Gene Symbol
SULT6B1
Entrez Gene ID
391365
Gene Name
sulfotransferase family 6B member 1
Gene Aliases
Chromosome Location
Chr. 2: 37167820 - 37188547 on Build GRCh38
UniGene ID
Hs.631892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001032377.1 NP_001027549.1 1 82
GenBank mRNA BC140797.1 - 1 82
BC144573.1 - 1 82

View Full Product Details