Gene Transcripts

Gene Symbol
IGLON5
Entrez Gene ID
402665
Gene Name
IgLON family member 5
Gene Aliases
Chromosome Location
Chr. 19: 51311848 - 51330848 on Build GRCh38
UniGene ID
Hs.546636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101372.1 NP_001094842.1 5 582

View Full Product Details