Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000243.2 | NP_000234.1 | 10 | 1947 | |
NM_001198536.1 | NP_001185465.1 | 9 | 1489 | ||
XM_017023236.1 | XP_016878725.1 | 10 | 2085 | ||
GenBank mRNA | AF018080.1 | - | 10 | 1948 | |
AK298660.1 | - | 10 | 1941 | ||
BC101511.1 | - | 10 | 1932 | 8069017 | |
BC101537.1 | - | 10 | 1932 | 8069043 | |
FJ785717.1 | - | 7 | 828 | ||
FJ785718.1 | - | 6 | 712 | ||
FJ785719.1 | - | 5 | 679 | ||
FJ785720.1 | - | 6 | 597 | ||
FJ785721.1 | - | 6 | 1312 | ||
FJ785722.1 | - | 9 | 1451 | ||
Y14441.1 | - | 10 | 1936 |