Gene Transcripts

Gene Symbol
LMOD2
Entrez Gene ID
442721
Gene Name
leiomodin 2
Gene Aliases
C-LMOD, CLMOD
Chromosome Location
Chr. 7: 123655807 - 123664093 on Build GRCh38
UniGene ID
Hs.592260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207163.1 NP_997046.1 2 1268
GenBank mRNA AK299389.1 - 2 1209
AK299434.1 - 2 1092
AK300698.1 - 3 1119

View Full Product Details