Gene Transcripts

Gene Symbol
NDUFA9
Entrez Gene ID
4704
Gene Name
NADH:ubiquinone oxidoreductase subunit A9
Gene Aliases
CC6, CI-39k, CI39k, NDUFS2L, SDR22E1
Chromosome Location
Chr. 12: 4649098 - 4687554 on Build GRCh38
UniGene ID
Hs.75227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005002.4 NP_004993.1 4 403
GenBank mRNA AF050641.1 - 4 393
AK291067.1 - 4 380
AK308588.1 - 4 384
BC003351.1 - 3 257
BC009311.2 - 4 384 4106931
BC015837.1 - 4 399 4293690
BC111546.1 - 4 375 5761015
CB110957.1 - 4 403
L04490.1 - 4 369

View Full Product Details