Gene Transcripts
- Gene Symbol
- NDUFB9
- Entrez Gene ID
- 4715
- Gene Name
- NADH:ubiquinone oxidoreductase subunit B9
- Gene Aliases
- B22, CI-B22, LYRM3, UQOR22
- Chromosome Location
- Chr. 8: 124539102 - 124549986 on Build GRCh38
- UniGene ID
- Hs.15977
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278645.1 |
NP_001265574.1
|
3, 4 |
449 |
|
NM_001278646.1 |
NP_001265575.1
|
3, 4 |
482 |
|
NM_001311168.1 |
NP_001298097.1
|
3, 4 |
451 |
|
NM_005005.2 |
NP_004996.1
|
3, 4 |
484 |
|
GenBank mRNA |
AF044956.1 |
- |
3, 4 |
478 |
|
AF067168.1 |
- |
3, 4 |
478 |
|
AF261090.1 |
- |
3, 4 |
483 |
|
AI907018.1 |
- |
3, 4 |
501 |
|
AK026538.1 |
- |
3, 4 |
467 |
|
AK313432.1 |
- |
3, 4 |
446 |
|
AL136714.1 |
- |
3, 4 |
454 |
|
BC007672.2 |
- |
3, 4 |
451 |
3160562 |
BG703246.1 |
- |
3, 4 |
411 |
|
BQ685604.1 |
- |
3, 4 |
434 |
|
CN410734.1 |
- |
3, 4 |
392 |
|
S82655.1 |
- |
2, 3 |
170 |
|
View Full Product Details