Gene Transcripts

Gene Symbol
CYHR1
Entrez Gene ID
50626
Gene Name
cysteine and histidine rich 1
Gene Aliases
CHRP
Chromosome Location
Chr. 8: 144449582 - 144465692 on Build GRCh38
UniGene ID
Hs.459379
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138496.1 NP_612505.1 2, 3 374
GenBank mRNA AK094514.1 - 2, 3 374

View Full Product Details