Gene Transcripts

Gene Symbol
GOLM1
Entrez Gene ID
51280
Gene Name
golgi membrane protein 1
Gene Aliases
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3
Chromosome Location
Chr. 9: 86026141 - 86100201 on Build GRCh38
UniGene ID
Hs.494337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016548.3 NP_057632.2 3 401
NM_177937.2 NP_808800.1 3 393
GenBank mRNA AF236056.1 - 3 342
AK027717.1 - 3 364
AK075542.1 - 3 335
AK075557.1 - 3 305
AK092943.1 - 3, 4 341
AY358593.1 - 3 306
BC001740.1 - 3 316 3351316
BQ216766.1 - 360
CR457201.1 - 2 192
DA116177.1 - 3 394
EU794616.1 - 3 360

View Full Product Details