Gene Transcripts
- Gene Symbol
- MTFP1
- Entrez Gene ID
- 51537
- Gene Name
- mitochondrial fission process 1
- Gene Aliases
- HSPC242, MTP18
- Chromosome Location
- Chr. 22: 30425623 - 30429054 on Build GRCh38
- UniGene ID
- Hs.713636
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003704.2 |
NP_001003704.1
|
1, 2 |
324 |
|
NM_016498.4 |
NP_057582.2
|
1, 2 |
324 |
|
GenBank mRNA |
AF060924.1 |
- |
1, 2 |
237 |
|
AK026104.1 |
- |
1, 2 |
192 |
|
BC001608.1 |
- |
1, 2 |
150 |
3542567 |
BC009300.2 |
- |
1, 2 |
182 |
4126237 |
BC014446.1 |
- |
1, 2 |
150 |
4855192 |
BC030989.2 |
- |
1, 2 |
211 |
4077757 |
BC038831.2 |
- |
1, 2 |
215 |
3908299 |
BC046132.1 |
- |
1, 2 |
114 |
4446034 |
BQ187590.1 |
- |
1, 2 |
106 |
|
CB107806.1 |
- |
1, 2 |
324 |
|
CB142469.1 |
- |
1, 2 |
215 |
|
CR456345.1 |
- |
1, 2 |
119 |
|
View Full Product Details