Gene Transcripts

Gene Symbol
LSM7
Entrez Gene ID
51690
Gene Name
LSM7 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
YNL147W
Chromosome Location
Chr. 19: 2321521 - 2328615 on Build GRCh38
UniGene ID
Hs.512610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016199.2 NP_057283.1 2 89
XM_017026869.1 XP_016882358.1 2 61
GenBank mRNA AF182293.1 - 1 49
BC018621.1 - 1 42 4855162
BI553911.1 - 2 94

View Full Product Details