Gene Transcripts
- Gene Symbol
- PI4KA
- Entrez Gene ID
- 5297
- Gene Name
- phosphatidylinositol 4-kinase alpha
- Gene Aliases
- PI4K-ALPHA, PIK4CA, PMGYCHA, pi4K230
- Chromosome Location
- Chr. 22: 20707691 - 20858812 on Build GRCh38
- UniGene ID
- Hs.529438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_058004.3 |
NP_477352.3
|
50, 51 |
5897 |
|
XM_005261634.1 |
XP_005261691.1
|
49, 50 |
5804 |
|
XM_005261635.1 |
XP_005261692.1
|
43, 44 |
5069 |
|
XM_017028829.1 |
XP_016884318.1
|
49, 50 |
5831 |
|
XM_017028830.1 |
XP_016884319.1
|
36, 37 |
4053 |
|
GenBank mRNA |
AB210002.1 |
- |
50, 51 |
5867 |
|
AF012872.1 |
- |
49, 50 |
5636 |
|
AK124692.1 |
- |
7, 8 |
1889 |
|
AK302424.1 |
- |
50, 51 |
5825 |
|
AK314781.1 |
- |
22, 23 |
2551 |
|
BC018120.1 |
- |
20, 21 |
2214 |
|
BC053654.1 |
- |
19, 20 |
2190 |
|
L36151.1 |
- |
18, 19 |
2183 |
|
- Gene Symbol
- PI4KAP2
- Entrez Gene ID
- 375133
- Gene Name
- phosphatidylinositol 4-kinase alpha pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 22: 21472998 - 21517491 on Build GRCh38
- UniGene ID
- Hs.448225
- Species
- Homo sapiens
View Full Product Details