Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318038.1 | NP_001304967.1 | 3 | 438 | |
NM_017436.5 | NP_059132.1 | 3 | 415 | ||
XM_005261644.2 | XP_005261701.1 | 3 | 720 | ||
XM_005261646.4 | XP_005261703.1 | 2 | 276 | ||
XM_005261647.2 | XP_005261704.1 | 3 | 250 | ||
XM_005261648.3 | XP_005261705.1 | 3 | 333 | ||
XM_006724265.3 | XP_006724328.1 | 3 | 661 | ||
XM_006724266.3 | XP_006724329.1 | 3 | 395 | ||
XM_011530233.2 | XP_011528535.1 | 2 | 602 | ||
XM_017028831.1 | XP_016884320.1 | 2 | 532 | ||
GenBank mRNA | AB037883.1 | - | 2 | 247 | |
AJ245581.1 | - | 1 | 114 | ||
AK312927.1 | - | 3 | 419 | ||
BC017068.2 | - | 3 | 334 | 3913851 | |
GU902275.1 | - | 2 | 205 | ||
GU902276.1 | - | 2 | 205 | ||
GU902277.1 | - | 3 | 277 | ||
KU178649.1 | - | 1 | 114 | ||
KU178650.1 | - | 1 | 114 |