Gene Transcripts

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
Chromosome Location
Chr. 11: 70078302 - 70189545 on Build GRCh38
UniGene ID
Hs.503074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018043.5 NP_060513.5 3, 4 845
NR_030691.1 3, 4 693
XM_006718602.2 XP_006718665.1 3, 4 561
XM_006718604.2 XP_006718667.1 3, 4 676
XM_006718605.2 XP_006718668.1 3, 4 688
XM_011545121.2 XP_011543423.1 3, 4 553
XM_011545123.2 XP_011543425.1 3, 4 553
XM_011545124.2 XP_011543426.1 3, 4 553
XM_011545125.2 XP_011543427.1 3, 4 553
XM_011545126.2 XP_011543428.1 3, 4 553
XM_011545127.2 XP_011543429.1 3, 4 553
XM_011545128.2 XP_011543430.1 3, 4 553
XM_011545129.2 XP_011543431.1 3, 4 468
XM_011545131.2 XP_011543433.1 3, 4 561
XM_017017956.1 XP_016873445.1 3, 4 553
XM_017017957.1 XP_016873446.1 3, 4 553
GenBank mRNA AB845669.1 - 3, 4 540
AK097619.1 - 3, 4 561
AK293088.1 - 3, 4 553
AL833582.1 - 3, 4 589
AY728143.1 - 3, 4 845
BC033036.2 - 3, 4 649 4823645
CF593428.1 - 3, 4 635

View Full Product Details