Gene Transcripts
- Gene Symbol
- TRIM36
- Entrez Gene ID
- 55521
- Gene Name
- tripartite motif containing 36
- Gene Aliases
- HAPRIN, RBCC728, RNF98
- Chromosome Location
- Chr. 5: 115124762 - 115180546 on Build GRCh38
- UniGene ID
- Hs.519514
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300752.1 |
NP_001287681.1
|
8, 9 |
2096 |
|
NM_001300759.1 |
NP_001287688.1
|
9, 10 |
2115 |
|
NM_018700.3 |
NP_061170.2
|
9, 10 |
2331 |
|
XM_017009621.1 |
XP_016865110.1
|
11, 12 |
2343 |
|
XM_017009622.1 |
XP_016865111.1
|
11, 12 |
3060 |
|
GenBank mRNA |
AB196558.1 |
- |
8, 9 |
1587 |
|
AJ272269.1 |
- |
9, 10 |
1948 |
|
AK058179.1 |
- |
4, 5 |
972 |
|
AK296389.1 |
- |
9, 10 |
2113 |
|
AK316422.1 |
- |
9, 10 |
2018 |
|
BC130334.1 |
- |
9, 10 |
2000 |
40146365 |
View Full Product Details