Gene Transcripts

Gene Symbol
PRTFDC1
Entrez Gene ID
56952
Gene Name
phosphoribosyl transferase domain containing 1
Gene Aliases
HHGP
Chromosome Location
Chr. 10: 24848607 - 24952644 on Build GRCh38
UniGene ID
Hs.405619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020200.6 NP_064585.1 8 634
GenBank mRNA AB209792.1 - 8 584
AF226056.1 - 8 582
AK021950.1 - 8 594
AK222674.1 - 8 593
BC008662.2 - 8 596 3871911

View Full Product Details