Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018070.2 | NP_001018080.1 | 5, 6 | 595 | |
NM_020441.2 | NP_065174.1 | 4, 5 | 573 | ||
GenBank mRNA | AK000860.1 | - | 4, 5 | 556 | |
AK295782.1 | - | 4, 5 | 533 | ||
AK309267.1 | - | 4 | 766 | ||
AK315399.1 | - | 4, 5 | 528 | ||
BC006449.1 | - | 4, 5 | 523 | 3958930 | |
BI767141.1 | - | 5, 6 | 571 |