Gene Transcripts

Gene Symbol
UGT2A2
Entrez Gene ID
574537
Gene Name
UDP glucuronosyltransferase family 2 member A2
Gene Aliases
Chromosome Location
Chr. 4: 69588417 - 69639642 on Build GRCh38
UniGene ID
Hs.225950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105677.2 NP_001099147.2 1 432
NM_001301233.1 NP_001288162.1 1 432
GenBank mRNA FJ664272.1 - 1 432
FJ664273.1 - 1 432

Gene Symbol
UGT2A1
Entrez Gene ID
10941
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Gene Aliases
UDPGT2A1
Chromosome Location
Chr. 4: 69588417 - 69653249 on Build GRCh38
UniGene ID
Hs.225950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252274.2 NP_001239203.2 3 1149
GenBank mRNA AK304249.1 - 3 1149

View Full Product Details