Gene Transcripts

Gene Symbol
PITPNM2
Entrez Gene ID
57605
Gene Name
phosphatidylinositol transfer protein membrane associated 2
Gene Aliases
NIR-3, NIR3, RDGB2, RDGBA2
Chromosome Location
Chr. 12: 122983480 - 123110489 on Build GRCh38
UniGene ID
Hs.272759
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300801.1 NP_001287730.1 4, 5 613
NM_020845.2 NP_065896.1 4, 5 552
XM_011538590.2 XP_011536892.1 4, 5 577
GenBank mRNA AB040890.2 - 5, 6 634
AF334585.1 - 3, 4 491
BC141813.1 - 4, 5 653

View Full Product Details