Gene Transcripts

Gene Symbol
ZSWIM6
Entrez Gene ID
57688
Gene Name
zinc finger SWIM-type containing 6
Gene Aliases
AFND
Chromosome Location
Chr. 5: 61332273 - 61546172 on Build GRCh38
UniGene ID
Hs.744939
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020928.1 NP_065979.1 11 2434
XM_017009677.1 XP_016865166.1 9 1385
GenBank mRNA AB046797.1 - 7 1020

View Full Product Details