Gene Transcripts

Gene Symbol
NPS
Entrez Gene ID
594857
Gene Name
neuropeptide S
Gene Aliases
Chromosome Location
Chr. 10: 127549349 - 127552671 on Build GRCh38
UniGene ID
Hs.643423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030013.1 NP_001025184.1 3 246

View Full Product Details