Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006509.3 | NP_006500.2 | 6, 7 | 886 | |
XM_005259127.3 | XP_005259184.1 | 5, 6 | 856 | ||
XM_005259128.2 | XP_005259185.1 | 6, 7 | 886 | ||
GenBank mRNA | AK290594.1 | - | 6, 7 | 793 | |
BC028013.1 | - | 6, 7 | 838 | 5215944 | |
M83221.1 | - | 6, 7 | 899 |