Gene Transcripts

Gene Symbol
RRAGC
Entrez Gene ID
64121
Gene Name
Ras related GTP binding C
Gene Aliases
GTR2, RAGC, TIB929
Chromosome Location
Chr. 1: 38838197 - 38859823 on Build GRCh38
UniGene ID
Hs.532461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271851.1 NP_001258780.1 5 910
NM_022157.3 NP_071440.1 5 1012
GenBank mRNA AF272035.1 - 5 840
AF323609.1 - 5 835
AK023373.1 - 1 40
AK298573.1 - 4 535
AK298788.1 - 5 806
AK315791.1 - 5 937
BC016668.1 - 5 857 4342448
CA449149.1 - 3 584

View Full Product Details