Gene Transcripts

Gene Symbol
KRTAP24-1
Entrez Gene ID
643803
Gene Name
keratin associated protein 24-1
Gene Aliases
KAP24.1
Chromosome Location
Chr. 21: 30281309 - 30282958 on Build GRCh38
UniGene ID
Hs.580883
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085455.2 NP_001078924.1 1 432
GenBank mRNA AB096935.1 - 1 432
AM238513.1 - 1 406

View Full Product Details