Gene Transcripts

Gene Symbol
SLIT1
Entrez Gene ID
6585
Gene Name
slit guidance ligand 1
Gene Aliases
MEGF4, SLIL1, SLIT-1, SLIT3
Chromosome Location
Chr. 10: 96998038 - 97185926 on Build GRCh38
UniGene ID
Hs.632082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003061.2 NP_003052.2 14 1574
GenBank mRNA AB011537.3 - 14 1574
AB017167.1 - 14 1554
AY029183.1 - 14 1301
BC146761.1 - 14 1574 9007200
BC146851.1 - 14 1487 9056779

View Full Product Details