Gene Transcripts

Gene Symbol
SPP1
Entrez Gene ID
6696
Gene Name
secreted phosphoprotein 1
Gene Aliases
BNSP, BSPI, ETA-1, OPN
Chromosome Location
Chr. 4: 87975650 - 87983411 on Build GRCh38
UniGene ID
Hs.313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000582.2 NP_000573.1 6 844
NM_001040058.1 NP_001035147.1 7 886
NM_001040060.1 NP_001035149.1 6 805
NM_001251829.1 NP_001238758.1 5 763
NM_001251830.1 NP_001238759.1 8 1075
XM_017008564.1 XP_016864053.1 5 1312
GenBank mRNA AB209987.1 - 4 598
AB469789.1 - 4 598
AF052124.1 - 6 766
AK057738.1 - 7 822
AK075463.1 - 8 756
AK290090.1 - 6 741
AK290104.1 - 7 823
AK295491.1 - 8 1010
AK296035.1 - 6 741
AK315461.1 - 6 774
BC007016.1 - 6 780 4052438
BC017387.1 - 7 826 3828885
BC022844.1 - 6 781 4284921
BC093033.1 - 6 797 5288027
BX648003.1 - 6 759
D28759.1 - 6 721
D28760.1 - 5 679
D28761.1 - 5 640
DQ839491.1 - 6 721
DQ846870.1 - 6 721
DQ846871.1 - 6 721
J04765.1 - 6 787
JF412666.1 - 7 657
JF412667.1 - 6 803
KR062183.1 - 8 1010
M83248.1 - 7 818
X13694.1 - 7 788

View Full Product Details