Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001051.4 | NP_001042.1 | 2 | 611 | |
NM_001278687.2 | NP_001265616.1 | 2 | 204 | ||
XM_005261721.4 | XP_005261778.1 | 2 | 104 | ||
XM_006724311.2 | XP_006724374.1 | 3 | 819 | ||
XM_011530349.2 | XP_011528651.1 | 7 | 954 | ||
XM_017028923.1 | XP_016884412.1 | 4 | 683 | ||
XM_017028924.1 | XP_016884413.1 | 4 | 1428 | ||
GenBank mRNA | AK291165.1 | - | 2 | 607 | |
BC042068.1 | - | 2 | 548 | ||
BC096829.1 | - | 1 | 32 | 6971896 | |
BU584980.1 | - | 2 | 480 | ||
CR456585.1 | - | 2 | 85 |