Gene Transcripts
- Gene Symbol
- VAMP1
- Entrez Gene ID
- 6843
- Gene Name
- vesicle associated membrane protein 1
- Gene Aliases
- SPAX1, SYB1, VAMP-1
- Chromosome Location
- Chr. 12: 6462237 - 6470899 on Build GRCh38
- UniGene ID
- Hs.20021
- Species
- Homo sapiens
- Gene Symbol
- TAPBPL
- Entrez Gene ID
- 55080
- Gene Name
- TAP binding protein like
- Gene Aliases
- TAPBP-R, TAPBPR
- Chromosome Location
- Chr. 12: 6451655 - 6472006 on Build GRCh38
- UniGene ID
- Hs.504597
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098035.1 |
- |
9 |
1736 |
|
View Full Product Details