Gene Transcripts

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
Chromosome Location
Chr. 2: 110637698 - 110678107 on Build GRCh38
UniGene ID
Hs.469649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278616.1 NP_001265545.1 19 2477
NM_001278617.1 NP_001265546.1 20 2537
NM_004336.4 NP_004327.1 20 2537
GenBank mRNA AF011387.1 - 12 1602
AF043294.2 - 20 2457
AF046078.1 - 20 2474
AF047471.1 - 20 2460
AF053305.1 - 20 2488
AK225317.1 - 20 2493
AK291221.1 - 20 2490
AK302418.1 - 19 2433
BC017038.1 - 20 2458
BC028201.1 - 20 2458 5205385

View Full Product Details