Gene Transcripts

Gene Symbol
TNNT2
Entrez Gene ID
7139
Gene Name
troponin T2, cardiac type
Gene Aliases
CMD1D, CMH2, CMPD2, LVNC6, RCM3, TnTC, cTnT
Chromosome Location
Chr. 1: 201359008 - 201377828 on Build GRCh38
UniGene ID
Hs.533613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000364.3 NP_000355.2 14 857
NM_001001430.2 NP_001001430.1 14 836
NM_001001431.2 NP_001001431.1 13 827
NM_001001432.2 NP_001001432.1 13 818
NM_001276345.1 NP_001263274.1 15 866
NM_001276346.1 NP_001263275.1 13 737
NM_001276347.1 NP_001263276.1 14 969
XM_006711508.3 XP_006711571.1 14 866
XM_006711509.3 XP_006711572.1 14 961
XM_011509938.2 XP_011508240.1 15 898
XM_011509939.1 XP_011508241.1 15 910
XM_011509940.2 XP_011508242.1 15 911
XM_011509941.2 XP_011508243.1 15 908
XM_011509942.2 XP_011508244.1 13 868
XM_011509943.2 XP_011508245.1 13 852
XM_011509944.2 XP_011508246.1 13 865
XM_011509946.1 XP_011508248.1 14 939
XM_017002216.1 XP_016857705.1 14 841
XM_017002217.1 XP_016857706.1 13 1020
GenBank mRNA AA865519.1 - 3 357
AK055533.1 - 4 1066
AK125236.1 - 13 1983
AK290621.1 - 14 1031
AK309493.1 - 8 670
AY277394.1 - 11 770
BC002653.2 - 13 793 3612968
L40162.1 - 14 797
S64668.1 - 13 756
X74819.1 - 13 794
X79861.1 - 13 783
X83743.1 - 4 214
X83744.1 - 3 211

View Full Product Details