Gene Transcripts

Gene Symbol
TPMT
Entrez Gene ID
7172
Gene Name
thiopurine S-methyltransferase
Gene Aliases
TPMTD
Chromosome Location
Chr. 6: 18128311 - 18155165 on Build GRCh38
UniGene ID
Hs.444319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000367.3 NP_000358.1 5 568
XM_011514839.2 XP_011513141.1 5 526
XM_017011241.1 XP_016866730.1 6 862
GenBank mRNA AF021876.1 - 4 391
AF021877.1 - 4 391
AK310420.1 - 5 462
BC005339.1 - 5 478 3829338
BC009596.1 - 5 477 4133029
BI598325.1 - 5 553
S62904.1 - 4 456
U12387.1 - 5 467

View Full Product Details