Gene Transcripts

Gene Symbol
MS4A18
Entrez Gene ID
728588
Gene Name
membrane spanning 4-domains A18
Gene Aliases
Chromosome Location
Chr. 11: 60722909 - 60744213 on Build GRCh38
UniGene ID
Hs.739039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001310141.1 NP_001297070.1 4, 5 1417
XM_011545420.2 XP_011543722.1 5, 6 2629
XM_011545421.2 XP_011543723.1 5, 6 2628

View Full Product Details