Gene Transcripts

Gene Symbol
PLIN4
Entrez Gene ID
729359
Gene Name
perilipin 4
Gene Aliases
KIAA1881, S3-12
Chromosome Location
Chr. 19: 4502180 - 4520285 on Build GRCh38
UniGene ID
Hs.591387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080400.1 NP_001073869.1 3 649
XM_006722866.2 XP_006722929.1 5 2610
XM_006722868.3 XP_006722931.1 5 2606
XM_011528233.2 XP_011526535.1 6 2792
XM_017027192.1 XP_016882681.1 6 2795
XM_017027193.1 XP_016882682.1 6 2742
XM_017027194.1 XP_016882683.1 6 2777
GenBank mRNA AB067468.1 - 425

View Full Product Details