Gene Transcripts

Gene Symbol
METTL12
Entrez Gene ID
751071
Gene Name
methyltransferase like 12
Gene Aliases
U99HG
Chromosome Location
Chr. 11: 62665252 - 62667451 on Build GRCh38
UniGene ID
Hs.640726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001043229.1 NP_001036694.1 3 528
XM_005274232.4 XP_005274289.1 2 897
GenBank mRNA AK297138.1 - 3 528
BC041359.1 - 2 980
BC147001.1 - 2 407 9056929
BC147011.1 - 2 407 9056939
BC171895.1 - 2 449
BX348415.2 - 1 95
BY796224.2 - 2 484

View Full Product Details