Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130721.1 | NP_001124193.1 | 4 | 364 | |
NM_024090.2 | NP_076995.1 | 3 | 567 | ||
XM_011532233.2 | XP_011530535.1 | 4 | 370 | ||
XM_011532234.2 | XP_011530536.1 | 4 | 438 | ||
XM_011532235.2 | XP_011530537.1 | 4 | 482 | ||
GenBank mRNA | AK027031.1 | - | 3 | 518 | |
AK074813.1 | - | 3 | 212 | ||
BC001305.1 | - | 4 | 351 | 3452423 | |
BP314858.1 | - | 4 | 364 |