Gene Transcripts

Gene Symbol
GDF5
Entrez Gene ID
8200
Gene Name
growth differentiation factor 5
Gene Aliases
BDA1C, BMP-14, BMP14, CDMP1, LAP-4, LAP4, OS5, SYM1B, SYNS2
Chromosome Location
Chr. 20: 35433347 - 35454746 on Build GRCh38
UniGene ID
Hs.1573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000557.4 NP_000548.2 1 313
NM_001319138.1 NP_001306067.1 3 555
XM_011529075.2 XP_011527377.1 3 445
GenBank mRNA AB593125.1 - 1 301
BC032495.1 - 3 555 5243109
U13660.1 - 1 313

View Full Product Details