Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142935.1 | NP_001136407.1 | 5 | 950 | |
NM_031300.3 | NP_112590.1 | 5 | 950 | ||
GenBank mRNA | AF114834.1 | - | 5 | 544 | |
AK057034.1 | - | 5 | 557 | ||
AK092842.1 | - | 1 | 267 | ||
AK222579.1 | - | 5 | 555 | ||
AK303397.1 | - | 5 | 557 | ||
AK316468.1 | - | 5 | 554 | ||
AL833959.1 | - | 2 | 1596 | ||
BC000745.2 | - | 5 | 539 | 2821596 | |
BC032586.1 | - | 5 | 554 | ||
BU620484.1 | - | 2 | 530 |