Gene Transcripts
- Gene Symbol
- GNPAT
- Entrez Gene ID
- 8443
- Gene Name
- glyceronephosphate O-acyltransferase
- Gene Aliases
- DAP-AT, DAPAT, DHAPAT, RCDP2
- Chromosome Location
- Chr. 1: 231241173 - 231277973 on Build GRCh38
- UniGene ID
- Hs.498028
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001316350.1 |
NP_001303279.1
|
9, 10 |
1541 |
|
NM_014236.3 |
NP_055051.1
|
10, 11 |
1724 |
|
XM_005273313.4 |
XP_005273370.1
|
10, 11 |
1684 |
|
XM_011544303.2 |
XP_011542605.1
|
11, 12 |
1773 |
|
XM_011544304.2 |
XP_011542606.1
|
10, 11 |
1592 |
|
GenBank mRNA |
AB209882.1 |
- |
10, 11 |
1629 |
|
AF035285.1 |
- |
4, 5 |
627 |
|
AF043937.1 |
- |
10, 11 |
1600 |
|
AJ002190.1 |
- |
10, 11 |
1675 |
|
AK222773.1 |
- |
10, 11 |
1680 |
|
AK223304.1 |
- |
10, 11 |
1683 |
|
AK297982.1 |
- |
9, 10 |
1502 |
|
AK311386.1 |
- |
9, 10 |
1374 |
|
AK314181.1 |
- |
10, 11 |
1687 |
|
BC000450.2 |
- |
10, 11 |
1571 |
2821393 |
View Full Product Details