Gene Transcripts

Gene Symbol
CASQ2
Entrez Gene ID
845
Gene Name
calsequestrin 2
Gene Aliases
PDIB2
Chromosome Location
Chr. 1: 115700003 - 115768805 on Build GRCh38
UniGene ID
Hs.57975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001232.3 NP_001223.2 8, 9 1100
GenBank mRNA AK129891.1 - 8, 9 1075
AK225959.1 - 8, 9 962
AK295502.1 - 6, 7 796
AK313041.1 - 8, 9 1046
BC022288.1 - 8, 9 1009 4289141
D55655.1 - 8, 9 945

View Full Product Details