Gene Transcripts

Gene Symbol
MFSD2A
Entrez Gene ID
84879
Gene Name
major facilitator superfamily domain containing 2A
Gene Aliases
MCPH15, MFSD2, NLS1
Chromosome Location
Chr. 1: 39955112 - 39969968 on Build GRCh38
UniGene ID
Hs.655177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136493.2 NP_001129965.1 2 309
NM_032793.4 NP_116182.2 2 309
NR_109896.1 2 309
XM_005271285.1 XP_005271342.1 2 273
XM_011542312.2 XP_011540614.1 2 276
GenBank mRNA AB073383.1 - 2 273
AF289609.1 - 2 265
AK074186.1 - 2 214
AK075183.1 - 2 276
AK093223.1 - 2 271
AK291540.1 - 2 271
AL834184.1 - 2 263
AY358636.1 - 2 290
BC011587.1 - 2 283 4646430
BC092414.1 - 2 251 30387073
BP321361.1 - 2 309
DC297497.1 - 2 309

View Full Product Details