Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005417.2 | NP_001005417.1 | 2 | 252 | |
NM_003780.4 | NP_003771.1 | 2 | 452 | ||
NM_030587.2 | NP_085076.2 | 2 | 253 | ||
XM_017002716.1 | XP_016858205.1 | 2 | 210 | ||
XM_017002717.1 | XP_016858206.1 | 2 | 413 | ||
GenBank mRNA | AB024434.1 | - | 1 | 98 | |
AF038660.1 | - | 1 | 139 | ||
AK095873.1 | - | 2 | 880 | ||
AK293418.1 | - | 2 | 202 | ||
AL535811.3 | - | 2 | 476 | ||
BC096821.1 | - | 2 | 220 | 30558762 | |
CD675528.1 | - | 2 | 260 | ||
DC386748.1 | - | 2 | 254 | ||
Y12510.1 | - | 1 | 90 |