Gene Transcripts

Gene Symbol
ZSWIM1
Entrez Gene ID
90204
Gene Name
zinc finger SWIM-type containing 1
Gene Aliases
C20orf162
Chromosome Location
Chr. 20: 45880920 - 45885266 on Build GRCh38
UniGene ID
Hs.517075
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080603.4 NP_542170.3 2 660
XM_005260610.4 XP_005260667.1 2 794
XM_005260611.4 XP_005260668.1 2 877
XM_011529100.2 XP_011527402.1 2 751
GenBank mRNA AK291304.1 - 2 648
BC001672.2 - 2 628 2988536
BC080525.1 - 2 628 4547006

View Full Product Details