Gene Transcripts

Gene Symbol
BRSK2
Entrez Gene ID
9024
Gene Name
BR serine/threonine kinase 2
Gene Aliases
C11orf7, PEN11B, SAD1, SADA, STK29
Chromosome Location
Chr. 11: 1389899 - 1462689 on Build GRCh38
UniGene ID
Hs.170819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256627.1 NP_001243556.1 18, 19 2323
NM_001256630.1 NP_001243559.1 18, 19 2328
NM_003957.3 NP_003948.2 18, 19 2323
XM_005253218.3 XP_005253275.1 18, 19 2035
XM_006718374.2 XP_006718437.1 19, 20 2101
XM_006718376.2 XP_006718439.1 19, 20 2013
XM_011520461.2 XP_011518763.1 18, 19 1947
XM_011520462.2 XP_011518764.1 18, 19 1947
XM_017018532.1 XP_016874021.1 19, 20 2379
XM_017018533.1 XP_016874022.1 19, 20 2467
XM_017018534.1 XP_016874023.1 19, 20 2379
XM_017018535.1 XP_016874024.1 18, 19 2313
XM_017018536.1 XP_016874025.1 18, 19 2401
XM_017018537.1 XP_016874026.1 19, 20 2379
XM_017018538.1 XP_016874027.1 14, 15 1924
GenBank mRNA AF020089.1 - 5, 6 551
AF533876.1 - 18, 19 2321
AF533879.1 - 18, 19 2321
AK131534.1 - 18, 19 2328
AY166857.1 - 18, 19 2323

View Full Product Details